Merck
Polypeptides of hepatitis B virus surface antigen produced by a hepatoma cell line.
Journal of virology (1979-12-01)
P L Marion, F H Salazar, J J Alexander, W S Robinson
PMID92575
ZUSAMMENFASSUNG
The PLC/PRF/5 cell line derived from a human hepatoma produces hepatitis B surface antigen (HBsAg) in 22-nm particles of the same buoyant density as those found in the serum of infected patients. The HBsAg particles from this cell line were labeled with [35S]methionine and purified, and the polypeptides were compared by sodium dodecyl sulfate-polyacrylamide gel electrophoresis with those of serum-derived particles. The two major polypeptides of serum-derived HBsAg particles (p20 and p23) were found in the same relative amounts in the particles from the cell line. The three smallest of the five minor components observed in HBsAg particles from serum were present in particles from the cell line. These polypeptides (p31, p36, and p43), as well as p20 and p23, were precipitated with anti-HBs-containing serum. The two largest polypeptides of serum particles (p49 and p66) were not detected in particles from these cells. When the PLC/PRF/5 HBsAg particles were radiolabeled with tritiated sugars, p23, and not p20, was found to contain radioactivity, indicating that the pattern of polypeptide glycosylation is similar to that of serum HBsAg. None of the other possible gene products of hepatitis B virus was detected in the PLC/PRF/5-derived HBsAg particles, in the cells, or in the cell supernatants.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
PLC/PRF/5, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.