Merck
Generation of a membrane potential by Lactococcus lactis through aerobic electron transport.
Journal of bacteriology (2007-05-15)
R J W Brooijmans, B Poolman, G K Schuurman-Wolters, W M de Vos, J Hugenholtz
ZUSAMMENFASSUNG
Lactococcus lactis, a facultative anaerobic lactic acid bacterium, is known to have an increased growth yield when grown aerobically in the presence of heme. We have now established the presence of a functional, proton motive force-generating electron transfer chain (ETC) in L. lactis under these conditions. Proton motive force generation in whole cells was measured using a fluorescent probe (3',3'-dipropylthiadicarbocyanine), which is sensitive to changes in membrane potential (Delta psi). Wild-type cells, grown aerobically in the presence of heme, generated a Delta psi even in the presence of the F(1)-F(o) ATPase inhibitor N,N'-dicyclohexylcarbodiimide, while a cytochrome bd-negative mutant strain (CydA Delta) did not. We also observed high oxygen consumption rates by membrane vesicles prepared from heme-grown cells, compared to CydA Delta cells, upon the addition of NADH. This demonstrates that NADH is an electron donor for the L. lactis ETC and demonstrates the presence of a membrane-bound NADH-dehydrogenase. Furthermore, we show that the functional respiratory chain is present throughout the exponential and late phases of growth.