Merck
  • Phase behavior of mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant
Phase behavior of mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant
Journal of Colloid and Interface Science (2007)
Tsuchiya, K., et al.