Merck
  • Design of grafted copper complex in mesoporous silica in defined coordination, hydrophobicity and confinement states
Design of grafted copper complex in mesoporous silica in defined coordination, hydrophobicity and confinement states
New. J. Chem. (2009)
Abry, S., et al.