Merck
  • Size-dependent self-assembly of submicron/nano beads-protein conjugates for construction of a protein nanoarray.
Size-dependent self-assembly of submicron/nano beads-protein conjugates for construction of a protein nanoarray.
Materials science & engineering. C, Materials for biological applications (2009-10-15)
Tremaine B Powell, Phat L Tran, Keesung Kim, Jeong-Yeol Yoon
ZUSAMMENFASSUNG
A protein nanoarray is created when submicro and nano beads, varying in their size and each conjugated with different proteins, self-assemble to specific locations depending on the diameter matching the surface electron beam patterns created. Protein binding is confirmed from the fluorescence attenuation of the beads upon antigen-antibody binding on the bead surface. This method, called size-dependent self-assembly, allows control of the location of each type of bead, and thus, control of the location of multiple proteins. It provides fast multi-component patterning with a high binding resolution, which can be detected using a fluorescent light microscope. This method is developed to be a simple stand-alone tool for analysis of protein interactions. In addition, it has the potential to be used in conjunction with other methods protein analysis methods, such as enzyme-linked immunosorbent assay (ELISA) and atomic force microscopy (AFM).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumborhydrid, powder, ≥98.0%
Sigma-Aldrich
IgG aus Mausserum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder