Merck
Fluorescence property of ZnO nanoparticles and the interaction with bromothymol blue.
Journal of fluorescence (2010-12-21)
Qiaoli Yue, Jinmei Cheng, Guang Li, Ke Zhang, Yanling Zhai, Lei Wang, Jifeng Liu
ZUSAMMENFASSUNG
We synthesized ZnO quantum dots (QDs) simply in alcoholic solution, and investigated the interaction between ZnO QDs and bromothymol blue. The structural, morphological, size and spectral properties of ZnO QDs were studied. It was found that ZnO QDs were spherical nanoparticles in the crystal structure, and the average diameter of ZnO QDs was about 4.8 nm. The excitation and emission peaks were located at 346 nm and 520 nm, respectively, which were obtained on a common fluorophotometer. The quantum yield of ZnO QDs was obtained by using quinine sulfate as a reference reagent. In addition, the fluorescence of ZnO QDs can be quenched by bromothymol blue, and the quenching mechanism was proposed in a dynamic quenching mode.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bromthymolblau, ACS reagent, Dye content 95 %
Sigma-Aldrich
Chinin -hemisulfat (Salz) Monohydrat, BioReagent, suitable for fluorescence, 99.0-101.0%