Merck
  • Monitoring of real changes of plasma membrane potential by diS-C(3)(3) fluorescence in yeast cell suspensions.
Monitoring of real changes of plasma membrane potential by diS-C(3)(3) fluorescence in yeast cell suspensions.
Journal of bioenergetics and biomembranes (2012-07-20)
Jaromír Plášek, Dana Gášková, Hella Lichtenberg-Fraté, Jost Ludwig, Milan Höfer
ZUSAMMENFASSUNG
The fluorescent dye 3,3'-dipropylthiadicarbocyanine, diS-C(3)(3), is a suitable probe to monitor real changes of plasma membrane potential in yeast cells which are too small for direct membrane potential measurements with microelectrodes. A method presented in this paper makes it possible to convert changes of equilibrium diS-C(3)(3) fluorescence spectra, measured in yeast cell suspensions under certain defined conditions, into underlying membrane potential differences, scaled in the units of millivolts. Spectral analysis of synchronously scanned diS-C(3)(3) fluorescence allows to assess the amount of dye accumulated in cells without otherwise necessary sample taking and following separation of cells from the medium. Moreover, membrane potential changes can be quantified without demanding calibration protocols. The applicability of this approach was demonstrated on the depolarization of Rhodotorula glutinis yeast cells upon acidification of cell suspensions and/or by increasing extracellular K(+) concentration.