Merck
  • Formulation of Tyloxapol niosomes for encapsulation, stabilization and dissolution of anti-tubercular drugs
Formulation of Tyloxapol niosomes for encapsulation, stabilization and dissolution of anti-tubercular drugs
Colloids and Surfaces. B, Biointerfaces (2013)
Mehta, S. and N. Jindal