Merck
  • Recent Developments in Porphyrin Separations Using Capillary Electrophoresis with Native Fluorescence Detection
Recent Developments in Porphyrin Separations Using Capillary Electrophoresis with Native Fluorescence Detection
Journal of Liquid Chromatography (1994)
Wu, N., et al.