Merck
  • Study on the distribution of binary mixed counterions in surfactant adsorbed films by total reflection XAFS measurements
Study on the distribution of binary mixed counterions in surfactant adsorbed films by total reflection XAFS measurements
Journal of Colloid and Interface Science (2012)
Y Imai et al.