Merck
  • Whole genome amplification and genetic analysis after extraction of proteins from dried blood spots.
Whole genome amplification and genetic analysis after extraction of proteins from dried blood spots.
Clinical chemistry (2007-05-23)
Mads V Hollegaard, Karina M Sørensen, Høgni K Petersen, Maria B Arnardottir, Bent Nørgaard-Pedersen, Poul Thorsen, David M Hougaard
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
GenomePlex® Kit zur Gesamtgenom-Amplifikation (GGA), Kit for whole genome amplification from a variety of DNA sources including FFPE tissue