Merck
  • Non-DNA-binding platinum anticancer agents: Cytotoxic activities of platinum-phosphato complexes towards human ovarian cancer cells.
Non-DNA-binding platinum anticancer agents: Cytotoxic activities of platinum-phosphato complexes towards human ovarian cancer cells.
Proceedings of the National Academy of Sciences of the United States of America (2008-11-21)
Rathindra N Bose, Leila Maurmann, Robert J Mishur, Linda Yasui, Shefalika Gupta, W Scott Grayburn, Heike Hofstetter, Tara Salley, Tara Milton
ZUSAMMENFASSUNG
DNA is believed to be the molecular target for the cytotoxic activities of platinum (Pt) anticancer drugs. We report here a class of platinum(II)- and platinum(IV)-pyrophosphato complexes that exhibit cytotoxicity comparable with and, in some cases, better than cisplatin in ovarian cell lines (A2780, A2780/C30, and CHO), yet they do not show any evidence of covalent binding to DNA. Moreover, some of these compounds are quite effective in cisplatin- and carboplatin-resistant cell line A2780/C30. The lack of DNA binding was demonstrated by the absence of a detectable Pt signal by atomic absorption spectroscopy using isolated DNA from human ovarian cells treated with a platinum(II)-pyrophosphato complex, (trans-1,2-cyclohexanediamine)(dihydrogen pyrophosphato) platinum(II), (pyrodach-2) and from NMR experiments using a variety of nucleotides including single- and double-stranded DNA. Furthermore, pyrodach-2 exhibited reduced cellular accumulations compared with cisplatin in cisplatin- and carboplatin-resistant human ovarian cells, yet the IC(50) value for the pyrophosphato complex was much less than that of cisplatin. Moreover, unlike cisplatin, pyrodach-2 treated cells overexpressed fas and fas-related transcription factors and some proapoptotic genes such as Bak and Bax. Data presented in this report collectively indicate that pyrodach-2 follows different cytotoxic mechanisms than does cisplatin. Unlike cisplatin, pyrodach-2 does not undergo aquation during 1 week and is quite soluble and stable in aqueous solutions. Results presented in this article represent a clear paradigm shift not only in expanding the molecular targets for Pt anticancer drugs but also in strategic development for more effective anticancer drugs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
SYBR® Green JumpStart Taq ReadyMix, for quantitative PCR, MgCI2 in buffer
Sigma-Aldrich
Referenzfarbstoff für quantitative PCR, 100 ×, solution