- Ruthenium-catalyzed reductive amination of allylic alcohols.
Ruthenium-catalyzed reductive amination of allylic alcohols.
Organic letters (2011-06-28)
Zeyneb Sahli, Basker Sundararaju, Mathieu Achard, Christian Bruneau
PMID21699264
ZUSAMMENFASSUNG
Straighforward access to various saturated amines from allylic alcohols and isostructural mixture can now be achieved in the presence of arene ruthenium catalyst featuring phosphinesulfonate ligand and a hydrogen donor.