- Isolation and characterization of two sequence-specific endonucleases from Anabaena variabilis.
Isolation and characterization of two sequence-specific endonucleases from Anabaena variabilis.
The Biochemical journal (1976-11-01)
K Murray, S G Hughes, J S Brown, S A Bruce
PMID11780
ZUSAMMENFASSUNG
Two endonucleases, AvaI and AvaII, were isolated from Anabaena variabilis on the basis of their ability to make a limited number of breaks at specific points in bacteriophage lambda DNA. Neither enzyme has cofactor requirements beyond Mg2+. Endonuclease AvaI makes eight breaks in the phage lambda chromosome at which the 5'-terminal sequence is pPy-C-G-N. AvaII endonuclease cuts phage lambda DNA more extensively, yielding fragments with the 5'-terminal sequence G-T-C-N or G-A-C-N. Neither enzyme generates cohesive ends.