- A specific endonuclease from Bacillus caldolyticus.
A specific endonuclease from Bacillus caldolyticus.
Nucleic acids research (1978-10-01)
A H Bingham, T Atkinson, D Sciaky, R J Roberts
PMID724493
ZUSAMMENFASSUNG
The purification and characterization of a new restriction endonuclease, BclI from the extreme thermophile Bacillus caldolyticus is reported. This enzyme recognizes the sequence : formula: (see text) and cleaves at the positions indicated by the arrows.