Merck
RsaI: a new sequence-specific endonuclease activity from Rhodopseudomonas sphaeroides.
Journal of bacteriology (1980-05-01)
S P Lynn, L K Cohen, S Kaplan, J F Gardner
ZUSAMMENFASSUNG
A new type II sequence-specific endonuclease, RsaI, has been identified from Rhodopseudomonas sphaeroides strain 28/5. An RsaI purification scheme that yields enzyme which is free of contaminating exonuclease and phosphatase activities after a single column fractionation has been developed. The enzyme recognized the tetranucleotide sequence 5'-GTAC-3' and cleaved between the T and A, thereby generating flush ends. RsaI should be extremely useful in deoxyribonucleic acid sequencing experiments.