Merck
  • Purification of recombinant human epidermal growth factor secreted from the methylotrophic yeast Hansenula polymorpha.
Purification of recombinant human epidermal growth factor secreted from the methylotrophic yeast Hansenula polymorpha.
Protein expression and purification (2002-01-29)
Joo-Hyung Heo, Hye Soon Won, Hyun Ah Kang, Sang-Ki Rhee, Bong Hyun Chung
ZUSAMMENFASSUNG
The gene encoding human epidermal growth factor (hEGF) was expressed as a fusion protein with the Saccharomyces cerevisiae-derived prepro alpha-factor leader in the methylotrophic yeast Hansenula polymorpha. The recombinant hEGF(1-53), when secreted by H. polymorpha, rapidly cleaved to hEGF(1-52) by carboxy-terminal proteolysis, resulting in the accumulation of C-terminal-truncated hEGF(1-52) in the culture medium. To solve this problem, we constructed a H. polymorpha mutant in which the KEX1 gene coding for carboxypeptidase ysc(alpha) was disrupted. The extent of C-terminal proteolysis of hEGF was significantly reduced when this kex1 disruptant was used as a host strain. After 24 h of shake-flask culture, most of the hEGF secreted by the kex1 disruptant remained intact, whereas more than 90% of the hEGF secreted by the wild-type was C-terminally cleaved. The recombinant hEGF was purified to >98% purity by two sequential steps of preparative scale anion exchange chromatography and reverse-phase HPLC. The authenticity of purified hEGF was confirmed by HPLC, N-terminal amino acid sequencing, and matrix-assisted laser desorption/ionization time-of-flight mass spectroscopy analyses.