Merck
  • Identification of a hexapeptide binding region in the nociceptin (ORL1) receptor by photo-affinity labelling with Ac-Arg-Bpa-Tyr-Arg-Trp-Arg-NH2.
Identification of a hexapeptide binding region in the nociceptin (ORL1) receptor by photo-affinity labelling with Ac-Arg-Bpa-Tyr-Arg-Trp-Arg-NH2.
Biochemical and biophysical research communications (2003-10-11)
Brice Bes, Jean-Claude Meunier
ZUSAMMENFASSUNG
The interaction of Ac-Arg-Tyr-Tyr-Arg-Trp-Arg-NH(2) (HP1), a high-affinity partial agonist of the opioid receptor like (ORL1) receptor, has been investigated using the photo-labile analogue [p-benzoyl-L-Phe (Bpa)(2)]-HP1. In recombinant CHO cells expressing the human ORL1 receptor, [Bpa(2)]-HP1 binds the receptor with high affinity (K; approximately 3nM) and is as potent as HP1 in stimulating GTPgammaS binding (50-60% of nociceptin maximal effect). UV irradiation at 365nm of the complex formed by the ORL1 receptor and radio-iodinated [Bpa(2)]-HP1 results in the irreversible labelling of a glycoprotein of M(r) approximately 66kDa, as determined by SDS-PAGE. Cyanogen bromide (CNBr) and enzymatic footprints of the photo-labelled receptor and an engineered receptor mutant (L113M), containing an additional CNBR cleavage site, allowed the photoreactive region to be identified as ORL1[107-113] at the C-terminal of TM helix II. In addition the presence of a disulphide bridge between Cysl23 and Cys200 has been confirmed biochemically.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Endoproteinase Asp-N aus Pseudomonas fragi mutant strain, suitable for protein sequencing, lyophilized powder