- SNOR and wheeze: the asthma enzyme?
SNOR and wheeze: the asthma enzyme?
Trends in molecular medicine (2005-10-11)
Edward M Henderson, Benjamin Gaston
PMID16214416
ZUSAMMENFASSUNG
Conventionally, asthma is defined as involving both airway inflammation and airway smooth muscle hyper-responsiveness. However, Que and coworkers have recently uncoupled these concepts, showing that mice lacking an S-nitrosothiol reductase have allergen-induced airway inflammation but do not have airway hyper-responsiveness. These data are consistent with recent clinical evidence that: (i) S-nitrosothiol signaling is abnormal in human asthma, (ii) nitric oxide in exhaled air might be only a biomarker for the metabolism of more physiologically relevant nitrogen oxides and (iii) the biochemical response to airway inflammation is central to asthma pathophysiology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Glutathion-Reduktase aus Backhefe (S. cerevisiae), ammonium sulfate suspension, 100-300 units/mg protein (biuret)