Merck
L-glutamate biosensor for estimation of the taste of tomato specimens.
Analytical and bioanalytical chemistry (2006-08-19)
Rasa Pauliukaite, Gleb Zhylyak, Daniel Citterio, Ursula E Spichiger-Keller
ZUSAMMENFASSUNG
An amperometric biosensor has been developed for measurement of Umami, or the taste based on the amount of L-glutamate, in tomato foods. The biosensor is based on an enzyme-mediator system in which L-glutamate oxidase is used for biochemical oxidation of L-glutamate and a tetrafulvalene-tetracyanoquinodimethane (TTF-TCNQ) paste, prepared from the mixture of TTF-TCNQ salt, graphite powder, and silicone oil, serves as the mediator. The limit of detection, calculated by use of a four-parameter logistic model, was 0.05 mmol L(-1), and the limit of quantification was 0.15 mmol L(-1). The correlation coefficient (R2) was 0.990 and the relative standard deviation was no more than 1% (n=5). The response time (tau (95)) was 20-50 s, depending on concentration. The repeatability of the sensor was better than 5% (n=10). The sensor developed was stable for more than ten days.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutamatoxidase aus Streptomyces sp., recombinant, expressed in E. coli, lyophilized powder, ≥5.0 units/mg solid