Merck
The catalytic mode of cysteine proteinases of papain (C1) family.
Bioresource technology (2006-09-26)
Leonidas G Theodorou, Joseph G Bieth, Emmanuel M Papamichael
ZUSAMMENFASSUNG
The Proton Inventory (PI) method has been applied in the hydrolysis of synthetic substrates by papain, chymopapain and stem bromelain, comparing also their corresponding pH-(k(cat)/K(m)) profiles, and it was found: (a) k(cat)/K(m)=k(1), and thus K(S)=k(2)/k(1) is a dynamic equilibrium constant, (b) bowed-downward PI for k(cat)/K(m) exhibiting large inverse SIE, and (c) linear PI exhibiting large normal SIE for K(S), k(2) and k(3). A novel finding of this work is that the association of substrates onto all three studied cysteine proteinases proceeds via a stepwise pathway, in contrast to purely concerted pathways found previously for both acylation and deacylation. A hydrogen bond, which seems more likely to be developed across a pK(a)-value close to 4.00, connecting [see text] (papain/chymopapain or bromelain numbering), constitutes another novelty of this work.