Merck
  • Identification of a 20-bp regulatory element of the Arabidopsis pyrophosphate:fructose-6-phosphate 1-phosphotransferase alpha2 gene that is essential for expression.
Identification of a 20-bp regulatory element of the Arabidopsis pyrophosphate:fructose-6-phosphate 1-phosphotransferase alpha2 gene that is essential for expression.
Plant cell reports (2007-01-06)
Hye-Min Lim, Jung-Il Cho, Sichul Lee, Man-Ho Cho, Seong Hee Bhoo, Gynheung An, Tae-Ryong Hahn, Jong-Seong Jeon
ZUSAMMENFASSUNG
Arabidopsis harbors two alpha and two beta genes of pyrophosphate:fructose-6-phosphate 1-phosphotransferase (PFP). The spatial expression patterns of the two AtPFPalpha genes were analyzed using transgenic plants containing a promoter::ss-glucuronidase (GUS) fusion construct. Whereas the AtPFPalpha1 promoter was found to be ubiquitously active in all tissues, the AtPFPalpha2 promoter is preferentially expressed in specific heterotrophic regions of the Arabidopsis plant such as the trichomes of leaves, cotyledon veins, roots, and the stamen and gynoecium of the flowers. Serial deletion analysis of the AtPFPalpha2 promoter identified a key regulatory element from nucleotides -194 to -175, CGAAAAAGGTAAGGGTATAT, which we have termed PFPalpha2 and which is essential for AtPFPalpha2 gene expression. Using a GUS fusion construct driven by this 20-bp sequence in conjunction with a -46 CaMV35S minimal promoter, we also demonstrate that PFPalpha2 is sufficient for normal AtPFPalpha2 expression. Hence, this element can not only be used to isolate essential DNA-binding protein(s) that control the expression of the carbon metabolic enzyme AtPFPalpha2, but has also the potential to be utilized in the production of useful compounds in a specific organ such as the leaf trichomes.