Merck
  • Activated thrombin activatable fibrinolysis inhibitor levels are associated with the risk of cardiovascular death in patients with coronary artery disease: the AtheroGene study.
Activated thrombin activatable fibrinolysis inhibitor levels are associated with the risk of cardiovascular death in patients with coronary artery disease: the AtheroGene study.
Journal of thrombosis and haemostasis : JTH (2008-11-20)
D A Tregouet, R Schnabel, M C Alessi, T Godefroy, P J Declerck, V Nicaud, T Munzel, C Bickel, H J Rupprecht, E Lubos, T Zeller, I Juhan-Vague, S Blankenberg, L Tiret, P E Morange
ZUSAMMENFASSUNG
Thrombin activatable fibrinolysis inhibitor (TAFI) attenuates fibrinolysis. Results on the association between TAFI levels and the risk of coronary artery disease (CAD) are inconsistent. We investigated the association between TAFI levels and the risk of cardiovascular events in CAD. 1668 individuals with angiographically proven CAD at baseline were followed for a median of 2.3 years, as part of the prospective AtheroGene cohort. Fifty-six deaths from cardiovascular (CV) causes and 35 non-fatal CV events were observed. At baseline, three TAFI measurements were available: one evaluating the total amount of TAFI (t-TAFI), one measuring the TAFIa/TAFIai amount, and the last the released activated peptide (TAFI-AP). TAFIa/TAFIai levels were associated with increased risk of CV death [hazard ratio (HR) for one tertile increase, 2.38 (1.56-3.63); P < 10(-4)]. This association remained significant after adjustment for conventional risk factors, CRP levels, white blood count and markers of thrombin generation and fibrinolysis [HR = 1.69 (1.07-2.67); P = 0.01]. In addition, CPB2 gene polymorphisms explained 12%, 6%, and 3% of t-TAFI, TAFIa/TAFIai and TAFI-AP levels, respectively, but none was associated with CV events. The amount of activated TAFI, measured by TAFIa/TAFIai ELISA, but not of the t-TAFI is independently associated with the risk of CV death.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Carboxypeptidase B aus Schweinepankreas, lyophilized powder
Sigma-Aldrich
Carboxypeptidase B aus meschlichem Pankreas, 50-55 units/mg protein carboxypeptidase B