Merck
  • Transglycosylation-based fluorescent labeling of 6-gala series glycolipids by endogalactosylceramidase.
Transglycosylation-based fluorescent labeling of 6-gala series glycolipids by endogalactosylceramidase.
Glycobiology (2009-04-25)
Yohei Ishibashi, Yusuke Nagamatsu, Sandra Meyer, Akihiro Imamura, Hideharu Ishida, Makoto Kiso, Nozomu Okino, Rudolf Geyer, Makoto Ito
ZUSAMMENFASSUNG
Although 6-gala series glycosphingolipids possessing R-Gal (alpha/beta) 1-6Gal beta 1-1'Cer have been found in some mollusks, pathogenic parasites, and fungi, their physiological functions and metabolic pathway are not fully understood. We described a novel method of detecting 6-gala series glyco- sphingolipids utilizing the specificity of endogalactosylceramidase (EGALC), which is capable of hydrolyzing 6-gala series glycosphingolipids to produce intact oligosaccharides and ceramides. EGALC catalyzes not only hydrolysis but also a transglycosylation reaction. In the latter reaction, EGALC transfers oligosaccharides from the glycosphingolipids to acceptors such as fluorescent 1-alkanols. Based on the transglycosylation reaction of EGALC, a specific, easy, fast, sensitive, and reproducible method of detecting 6-gala series glycosphingolipids was developed using NBD-pentanol as an acceptor. The fluorescent products, NBD-pentanol-conjugated 6-gala oligosaccharides, were separated and detected by TLC or HPLC with a fluorescent detector. Moreover, it was revealed that as well as glycosphingolipids, a glycoglycerolipid, digalactosyldiacylglycerol, was utilized by EGALC as a donor substrate. This method was successfully applied to detect 6-gala series glycosphingolipids in a fungus, Rhizopus oryzae, and a parasite, Taenia crassiceps. The method would be useful for studying glycosphingolipids and galactosyl glycerolipids which share the Gal (alpha/beta) 1-6Gal structure.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Endoglycoceramidase II from Rhodococcus sp., aqueous solution