Merck
  • Cholesterol oxidase ChoD is not a critical enzyme accounting for oxidation of sterols to 3-keto-4-ene steroids in fast-growing Mycobacterium sp. VKM Ac-1815D.
Cholesterol oxidase ChoD is not a critical enzyme accounting for oxidation of sterols to 3-keto-4-ene steroids in fast-growing Mycobacterium sp. VKM Ac-1815D.
The Journal of steroid biochemistry and molecular biology (2011-10-22)
Tatyana V Ivashina, Vera M Nikolayeva, Dmitry V Dovbnya, Marina V Donova
ZUSAMMENFASSUNG
Fast-growing strain of Mycobacterium sp. VKM Ac-1815D is capable of effective oxidizing of sterols (phytosterol, cholesterol, ergosterol) to androstenedione and other valuable 3-oxo-steroids. To elucidate the role of cholesterol oxidase in sterol catabolism by the strain, the choD gene has been cloned and sequenced. The deduced gene product (M(r) 63.5kDa) showed homologies over its entire length to a large number of proteins belonging to the InterPro-family EPR006076, which includes various FAD dependent oxidoreductases. The expression of choD in Escherichia coli was shown to result in the synthesis of membrane associated cholesterol oxidase. In addition to cholesterol, the enzyme oxidized β-sitosterol, dehydroepiandrosterone, ergosterol, pregnenolone, and lithocholic acid. Knock-out of choD in Mycobacterium sp. VKM Ac-1815D strain was obtained by the gene replacement technique. The mutant strain transformed sitosterol forming exclusively 3-keto-4-ene steroids with androstenedione as a major product, thus evidencing that choD knock out did not abrogate sterol A-ring oxidation. The results indicated that ChoD is not a critical enzyme responsible for modification of 3β-hydroxy-5-ene- to 3-keto-4-ene steroids in Mycobacterium sp. VKM Ac-1815D. Article from a special issue on steroids and microorganisms.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterol Oxidase from Brevibacterium, lyophilized powder, ≥50 units/mg protein, recombinant, expressed in E. coli
Sigma-Aldrich
Cholesterin-Oxidase aus Streptomyces sp., lyophilized powder, ≥20 units/mg protein
Sigma-Aldrich
Cholesterol Oxidase microbial, recombinant, lyophilized powder, ≥10 units/mg protein
Sigma-Aldrich
Cholesterol Oxidase from microorganisms, aqueous solution, ≥30 units/mg protein (biuret)