Merck
  • Effect of Ca2+ channel block on glycerol metabolism in Dunaliella salina under hypoosmotic and hyperosmotic stresses.
Effect of Ca2+ channel block on glycerol metabolism in Dunaliella salina under hypoosmotic and hyperosmotic stresses.
PloS one (2011-12-24)
Hui Chen, Shan-Li Chen, Jian-Guo Jiang
ZUSAMMENFASSUNG
The effect of Ca(2+) channel blockers on cytosolic Ca(2+) levels and the role of Ca(2+) in glycerol metabolism of Dunaliella salina under hypoosmotic or hyperosmotic stress were investigated using the confocal laser scanning microscope (CLSM). Results showed that intracellular Ca(2+) concentration increased rapidly when extracellular salinity suddenly decreased or increased, but the increase could be inhibited by pretreatment of Ca(2+) channel blockers LaCl(3), verapamil or ruthenium red. The changes of glycerol content and G3pdh activity in D. salina to respect to hypoosmotic or hyperosmotic stress were also inhibited in different degrees by pretreatment of Ca(2+) channel blockers, indicating that the influx of Ca(2+) via Ca(2+) channels are required for the transduction of osmotic signal to regulate osmotic responses of D. salina to the changes of salinity. Differences of the three blockers in block effect suggested that they may act on different channels or had different action sites, including influx of Ca(2+) from the extracellular space via Ca(2+) channels localized in the plasma membrane or from intracellular calcium store via the mitochondrial. Other Ca(2+)-mediated or non-Ca(2+)-mediated osmotic signal pathway may exist in Dunaliella in response to hypoosmotic and hyperosmotic stresses.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-Glycerophosphatdehydrogenase-Triosephosphatisomerase aus Kaninchenmuskel, Type III, ammonium sulfate suspension
Sigma-Aldrich
α-Glycerophosphat-Dehydrogenase aus Kaninchenmuskel, Type I, ammonium sulfate suspension, 100-300 units/mg protein
Sigma-Aldrich
α-Glycerophosphat-Dehydrogenase aus Kaninchenmuskel, Type X, lyophilized powder, ≥100 units/mg protein