Merck
  • Heterologous expression of manganese peroxidase in Aspergillus niger and its effect on phenanthrene removal from soil.
Heterologous expression of manganese peroxidase in Aspergillus niger and its effect on phenanthrene removal from soil.
Journal of molecular microbiology and biotechnology (2012-01-31)
Diana V Cortés-Espinosa, Ángel E Absalón, Noé Sanchez, Octavio Loera, Refugio Rodríguez-Vázquez, Francisco J Fernández
ZUSAMMENFASSUNG
A strain of Aspergillus niger, previously isolated from sugarcane bagasse because of its capacity to degrade phenanthrene in soil by solid culture, was used to express a manganese peroxidase gene (mnp1) from Phanerochaete chrysosporium, aiming at increasing its polycyclic aromatic hydrocarbons degradation capacity. Transformants were selected based on their resistance to hygromycin B and the discoloration induced on Poly R-478 dye by the peroxidase activity. The recombinant A. niger SBC2-T3 strain developed MnP activity and was able to remove 95% of the initial phenanthrene (400 ppm) from a microcosm soil system after 17 days, whereas the wild strain removed 72% under the same conditions. Transformation success was confirmed by PCR amplification using gene-specific primers, and a single fragment (1,348 bp long, as expected) of the recombinant mnp1 was amplified in the DNA from transformants, which was absent from the parental strain.