- Improvement of the transcriptional strength of baculovirus very late polyhedrin promoter by repeating its untranslated leader sequences and coexpression with the primary transactivator.
Improvement of the transcriptional strength of baculovirus very late polyhedrin promoter by repeating its untranslated leader sequences and coexpression with the primary transactivator.
Journal of bioscience and bioengineering (2012-02-09)
Tatsuya Kato, Suganthi Lavender Manohar, Shin Kanamasa, Makoto Ogata, Enoch Y Park
PMID22309650
ZUSAMMENFASSUNG
Modified polyhedrin promoter (Ppolh) was designed by repeating burst sequences (BSs) and adopted to overexpress rat α2,6-sialyltransferase (ST6Gal I) in silkworm. Modified Ppolh of five BSs with VLF-1 coexpression yielded 2.9 U/ml ST6Gal I activity and 32.5 mU/mg specific activity, which was 1.7- and 2.3-fold higher, respectively compared to Ppolh.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
α-2,6-Sialyltransferase from Photobacterium damsela, recombinant, expressed in E. coli BL21, ≥5 units/mg protein