Merck
Integrated photosystem II-based photo-bioelectrochemical cells.
Nature communications (2012-03-15)
Omer Yehezkeli, Ran Tel-Vered, Julian Wasserman, Alexander Trifonov, Dorit Michaeli, Rachel Nechushtai, Itamar Willner
ZUSAMMENFASSUNG
Photosynthesis is a sustainable process that converts light energy into chemical energy. Substantial research efforts are directed towards the application of the photosynthetic reaction centres, photosystems I and II, as active components for the light-induced generation of electrical power or fuel products. Nonetheless, no integrated photo-bioelectrochemical device that produces electrical power, upon irradiation of an aqueous solution that includes two inter-connected electrodes is known. Here we report the assembly of photobiofuel cells that generate electricity upon irradiation of biomaterial-functionalized electrodes in aqueous solutions. The cells are composed of electrically contacted photosystem II-functionalized photoanodes and an electrically wired bilirubin oxidase/carbon nanotubes-modified cathode. Illumination of the photoanodes yields the oxidation of water to O(2) and the transfer of electrons through the external circuit to the cathode, where O(2) is re-reduced to water.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Bilirubinoxidase aus Myrothecium verrucaria, lyophilized powder, 15-65 units/mg protein