Merck
  • Purification, characterization and decolorization of bilirubin oxidase from Myrothecium verrucaria 3.2190.
Purification, characterization and decolorization of bilirubin oxidase from Myrothecium verrucaria 3.2190.
Fungal biology (2012-08-07)
Xu Han, Min Zhao, Lei Lu, Yanhong Liu
ZUSAMMENFASSUNG
Myrothecium verrucaria 3.2190 is a nonligninolytic fungus that produces bilirubin oxidase. Both M. verrucaria and the extracellular bilirubin oxidase were tested for their ability to decolorize indigo carmine. The biosorption and biodegradation of the dye were detected during the process of decolorization; more than 98% decolorization efficiency was achieved after 7 days at 26°C. Additionally, the crude bilirubin oxidase can efficiently decolorize indigo carmine at 30°C~50°C, pH 5.5~9.5 with dye concentrations of 50 mg l(-1)~200 mg l(-1). Bilirubin oxidase was purified and visualized as a single band on native polyacrylamide gel electrophoresis (PAGE). Several enzymatic properties of the purified enzyme were investigated. Moreover, the identity of the purified bilirubin oxidase (BOD) was confirmed by matrix assisted laser desorption ionisation time-of-flight mass spectrometry (MALDI-TOF-MS). These results demonstrate that the purified bilirubin oxidase in M. verrucaria strain has potential application in dye effluent decolorization.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Indigokarmin, suitable for microscopy
Supelco
Indigokarmin, analytical standard
Sigma-Aldrich
Indigokarmin, certified by the BSC, Dye content 85 %
Sigma-Aldrich
Bilirubinoxidase aus Myrothecium verrucaria, lyophilized powder, 15-65 units/mg protein