- A chemically programmed antibody is a long-lasting and potent inhibitor of influenza neuraminidase.
A chemically programmed antibody is a long-lasting and potent inhibitor of influenza neuraminidase.
Chembiochem : a European journal of chemical biology (2012-09-12)
Masahiko Hayakawa, Narihiro Toda, Nancy Carrillo, Natalie J Thornburg, James E Crowe, Carlos F Barbas
PMID22965667
ZUSAMMENFASSUNG
Programming an anti-flu strategy: A new and potent neuraminidase inhibitor that maintains long-term systemic exposure of an antibody and the therapeutic activity of the neuraminadase inhibitor zanamivir has been created. This strategy could provide a promising new class of influenza A drugs for therapy and prophylaxis, and validates enzyme inhibitors as programming agents in synthetic immunology.