Merck
  • [A redox process of ethanethiol group increases formation of beta-sheet structure and amyloid fibrils of wild type human prion protein].
[A redox process of ethanethiol group increases formation of beta-sheet structure and amyloid fibrils of wild type human prion protein].
Bing du xue bao = Chinese journal of virology (2012-09-18)
Han Sun, Qi Shi, Shao-Bin Wang, Fei Guo, Wu-Ling Xie, Cao Chen, Cun-Qi Liu, Xiao-Ping Dong
ZUSAMMENFASSUNG
In order to explore the potential influences of the disulfide bridge on the physical and chemical properties of PrP protein, the expressed recombinant human wild-type PrP protein was purified for using in an established redox process for the reduction and oxidation of the ethanethiol group within PrP. Sedimentation tests illustrated that redox process remarkably promoted the aggregation of recombinant PrP. Thioflavin T binding assay revealed an enhanced fibrillization of the recombinant human PrP after redox process. Far-UV circular dichroism demonstrated that the PrP treated with redox process showed a significant p-sheet rich structure. Furthermore, PrP-specific Western blot identified that the recombinant PrP after redox possessed stronger proteinase K-resistance. Those data indicates that the formation of the disulfide bridge induces the alteration of the secondary structure and enhances the progresses of aggregation and fibrillization of PrP protein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteinase K aus Tritirachium album, lyophilized powder, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K aus Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K aus Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL
Sigma-Aldrich
Ethanethiol, 97%
Sigma-Aldrich
Proteinase K aus Tritirachium album, ≥3.0 unit/mg solid, lyophilized powder
Sigma-Aldrich
Natrium-ethanthiolat, technical, ~90% (RT)
Sigma-Aldrich
Proteinase K aus Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution
Sigma-Aldrich
Natrium-ethanthiolat, technical grade, 80%
Sigma-Aldrich
Ethanethiol, ≥97%