- Evaluation of six commercial DNA extraction kits for recovery of Burkholderia pseudomallei DNA.
Evaluation of six commercial DNA extraction kits for recovery of Burkholderia pseudomallei DNA.
Journal of microbiological methods (2012-10-02)
Maria Angela de Mello Marques, Pia Zimmermann, Ute Messelhäußer, Andreas Sing
PMID23022447
ZUSAMMENFASSUNG
Six commercially available DNA extraction kits, as well as thermal lysis and proteinase K DNA extraction were evaluated regarding bacterial inactivation, DNA yield and purity, and their use in a Burkholderia pseudomallei real-time PCR. While all methods successfully inactivated the bacteria, by measuring DNA purity and the level of detection by real-time PCR, the proteinase K method was the most sensitive.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteinase K aus Tritirachium album, lyophilized powder, Molecular Biology, BioUltra, ≥30 units/mg protein
Sigma-Aldrich
Proteinase K aus Tritirachium album, buffered aqueous glycerol solution, Molecular Biology, ≥800 units/mL
Sigma-Aldrich
Proteinase K aus Tritirachium album, ≥500 units/mL, buffered aqueous glycerol solution