Merck
  • Structural and molecular insights into the mechanism of action of human angiogenin-ALS variants in neurons.
Structural and molecular insights into the mechanism of action of human angiogenin-ALS variants in neurons.
Nature communications (2012-10-11)
Nethaji Thiyagarajan, Ross Ferguson, Vasanta Subramanian, K Ravi Acharya
ZUSAMMENFASSUNG
Mutations in angiogenin (ANG), a member of the ribonuclease A superfamily, are associated with amyotrophic lateral sclerosis (ALS; sporadic and familial) and Parkinson's disease. We have previously shown that ANG is expressed in neurons during neuro-ectodermal differentiation, and that it has both neurotrophic and neuroprotective functions. Here we report the atomic resolution structure of native ANG and 11 ANG-ALS variants. We correlate the structural changes to the effects on neuronal survival and the ability to induce stress granules in neuronal cell lines. ANG-ALS variants that affect the structure of the catalytic site and either decrease or increase the RNase activity affect neuronal survival. Neuronal cell lines expressing the ANG-ALS variants also lack the ability to form stress granules. Our structure-function studies on these ANG-ALS variants are the first to provide insights into the cellular and molecular mechanisms underlying their role in ALS.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease B aus Rinderpankreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein