- [Investigation of functional groups of Cryptococcus albidus alpha-L-rhamnosidase].
[Investigation of functional groups of Cryptococcus albidus alpha-L-rhamnosidase].
Mikrobiolohichnyi zhurnal (Kiev, Ukraine : 1993) (2012-10-24)
O V Gudzenko, L D Varbanets'
PMID23088096
ZUSAMMENFASSUNG
The effect of cations, anions and specific chemical reagents: 1-[3-(dimethylamino)propyl]-3-ethylcarbodimide methiodide, EDTA, o-phenantroline, dithiotreitol, L-cysteine, beta-mercaptoethanol, p-chlormercurybenzoate (p-ChMB), N-ethylmaleimide on the alpha-L-rhamnosidase activity of Cryptococcus albidus has been investigated. The essential role of Ag+ which inhibits the alpha-L-rhamnosidase activity by 72.5% was shown. Rhamnose at 1-5 mM protect the enzyme from the negative effect of Ag(+). It was expected that carboxyl group of C-terminal aminoacid and imidazole group of histidine would participate in the catalytic action of alpha-L-rhamnosidase on the basis of inhibition and kinetic analysis.