Merck
  • The coagulopathy and thrombotic risk associated with L-asparaginase treatment in adults with acute lymphoblastic leukaemia.
The coagulopathy and thrombotic risk associated with L-asparaginase treatment in adults with acute lymphoblastic leukaemia.
Leukemia (2012-10-27)
E Truelove, A K Fielding, B J Hunt
ZUSAMMENFASSUNG
The dramatic improvements seen in the outcome of paediatric patients with acute lymphoblastic leukaemia (ALL) have led to increasing incorporation of L-asparaginase (L-Asp) in adult treatment protocols. However, its use is associated with a disruption in the physiological balance between haemostatic and anticoagulant pathways, with the predominant clinical manifestation being thrombosis. Although L-Asp therapy is known to be associated with an acquired deficiency of antithrombin (AT), the concurrent depletion of fibrinogen and other haemostatic proteins means that the precise mechanism of thrombosis remains to be defined. In vitro coagulation assays are often prolonged but thrombosis rather than haemorrhage is the primary concern. Management of thrombotic events in these patients is based around agents that rely on AT for their anticoagulant effect, even though it is usually depleted. There is currently only limited evidence supporting the use of AT concentrates in either primary prevention or management following an established event. Evidence-based guidelines for prevention and management strategies are lacking.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Asparaginase aus E. coli, lyophilized powder, 100-300 units/mg protein (biuret)