Merck
The anti-obesity effect of natural vanadium-containing Jeju ground water.
Biological trace element research (2012-12-04)
Seon-Joo Park, Cha-Kyung Youn, Jin Won Hyun, Ho Jin You
ZUSAMMENFASSUNG
This study investigated the anti-obesity effects of Jeju ground water containing the vanadium components S1 (8.0 ± 0.9 μg/l) and S3 (26.0 ± 2.09 μg/l) on the differentiation of 3 T3-L1 preadipocytes and obesity in mice that were fed a high-fat diet (HFD). The 3 T3-L1 preadipocyte cells were cultured and differentiated in media consisting of Jeju ground water (S1, S3) or deionized water (DW) containing dexamethasone, isobutylmethylxanthine, and insulin. Oil Red O staining showed that lipid accumulation was attenuated in adipocyte cells treated with Jeju ground water. S3 significantly decreased peroxisome-activated receptor γ and CCAAT-enhancer-binding protein α mRNA expression levels, which play major roles in the transcriptional control of adipogenesis, compared to DW. Furthermore, mRNA expression levels of targeted genes, such as adipocyte fatty acid, lipoprotein lipase, and leptin, were decreased by S3 treatment compared with the control group. In mice with HFD-induced obesity, Jeju ground water decreased HFD-induced body weight gain and reduced total cholesterol, triglyceride, and glucose levels in the plasma compared to control mice. Taken together, Jeju ground water inhibits preadipocyte differentiation and adipogenesis in obesity animal models.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ölrot O -Lösung, 0.5% in isopropanol
Sigma-Aldrich
Ölrot O, certified by the BSC
Sigma-Aldrich
Lipoprotein-Lipase aus Kuhmilch, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)
Sigma-Aldrich
Lipoprotein-Lipase aus Pseudomonas sp., lyophilized, powder, ≥1200 U/mg
Sigma-Aldrich
Ölrot O -Lösung, 0.5% in propylene glycol
Sigma-Aldrich
Vanadium, powder, −325 mesh, 99.5% trace metals basis
Supelco
Ölrot O, BioReagent, suitable for electrophoresis
Sigma-Aldrich
Lipoprotein-Lipase aus Pseudomonas sp., lyophilized powder, ≥50,000 units/mg solid
Sigma-Aldrich
Vanadium, foil, thickness 0.127 mm, 99.7% trace metals basis