Merck
Reversible inactivation of dihydrolipoamide dehydrogenase by mitochondrial hydrogen peroxide.
Free radical research (2012-12-05)
Liang-Jun Yan, Nathalie Sumien, Nopporn Thangthaeng, Michael J Forster
ZUSAMMENFASSUNG
Under oxidative stress conditions, mitochondria are the major site for cellular production of reactive oxygen species (ROS) such as superoxide anion and H2O2 that can attack numerous mitochondrial proteins including dihydrolipoamide dehydrogenase (DLDH). While DLDH is known to be vulnerable to oxidative inactivation, the mechanisms have not been clearly elucidated. The present study was therefore designed to investigate the mechanisms of DLDH oxidative inactivation by mitochondrial reactive oxygen species (ROS). Mitochondria, isolated from rat brain, were incubated with mitochondrial respiratory substrates such as pyruvate/malate or succinate in the presence of electron transport chain inhibitors such as rotenone or antimycin A. This is followed by enzyme activity assay and gel-based proteomic analysis. The present study also examined whether ROS-induced DLDH oxidative inactivation could be reversed by reducing reagents such as DTT, cysteine, and glutathione. Results show that DLDH could only be inactivated by complex III- but not complex I-derived ROS; and the accompanying loss of activity due to the inactivation could be restored by cysteine and glutathione, indicating that DLDH oxidative inactivation by complex III-derived ROS was a reversible process. Further studies using catalase indicate that it was H2O2 instead of superoxide anion that was responsible for DLDH inactivation. Moreover, using sulfenic acid-specific labeling techniques in conjunction with two-dimensional Western blot analysis, we show that protein sulfenic acid formation (also known as sulfenation) was associated with the loss of DLDH enzymatic activity observed under our experimental conditions. Additionally, such oxidative modification was shown to be associated with preventing DLDH from further inactivation by the thiol-reactive reagent N-ethylmaleimide. Taken together, the present study provides insights into the mechanisms of DLDH oxidative inactivation by mitochondrial H2O2.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Ethylmaleimid, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimid, crystalline, ≥98% (HPLC)
Sigma-Aldrich
DL-Apfelsäure, ReagentPlus®, ≥99%
Sigma-Aldrich
N-Ethylmaleimid, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
DL-Apfelsäure, 99%
Sigma-Aldrich
Diaphorase aus Clostridium kluyveri, lyophilized powder, 3.0-20.0 units/mg protein (biuret)
Sigma-Aldrich
DL-Apfelsäure, ≥98% (capillary GC)
Sigma-Aldrich
N-Ethylmaleimid, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Diaphorase from Clostridium kluyveri, recombinant, expressed in E. coli, Animal-component free
Supelco
D-Apfelsäure, analytical standard
Sigma-Aldrich
L-(−)-Maleinsäure Dinatriumsalz, ≥95% (titration)
Sigma-Aldrich
Apfelsäure, meets USP/NF testing specifications
Sigma-Aldrich
DL-Apfelsäure, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
D(+)-Äpfelsäure, unnatural form, ≥97.0% (T)