Merck
Fatal invasive aspergillosis of the larynx.
European journal of haematology (2012-12-12)
Mark S Williams, Nisha Ali, Daisuke Nonaka, Adrian J Bloor, Tim C P Somervaille
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Asparaginase aus E. coli, lyophilized powder, 100-300 units/mg protein (biuret)