Merck
Surface display of a redox enzyme and its site-specific wiring to gold electrodes.
Journal of the American Chemical Society (2012-12-13)
Liron Amir, Stewart A Carnally, Josep Rayo, Shaked Rosenne, Sarit Melamed Yerushalmi, Orr Schlesinger, Michael M Meijler, Lital Alfonta
ZUSAMMENFASSUNG
The generation of a current through interaction between bacteria and electrodes has been explored by various methods. We demonstrate the attachment of living bacteria through a surface displayed redox enzyme, alcohol dehydrogenase II. The unnatural amino acid para-azido-L-phenylalanine was incorporated into a specific site of the displayed enzyme, facilitating electron transfer between the enzyme and an electrode. In order to attach the bacteria carrying the surface displayed enzyme to a surface, a linker containing an alkyne and a thiol moiety on opposite ends was synthesized and attached to the dehydrogenase site specifically through a copper(I)-catalyzed azide-alkyne cycloaddition reaction. Using this approach we were able to covalently link bacteria to gold-coated surfaces and to gold nanoparticles, while maintaining viability and catalytic activity. We show the performance of a biofuel cell using these modified bacteria at the anode, which resulted in site-specific dependent fuel cell performance for at least a week. This is the first example of site-specific attachment of a true living biohybrid to inorganic material.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Alcohol-Dehydrogenase aus Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Alcohol-Dehydrogenase aus Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Alcohol-Dehydrogenase aus Saccharomyces cerevisiae
Sigma-Aldrich
Alkoholdehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg