Merck
Erythrocyte adenosine deaminase: diagnostic value for Diamond-Blackfan anaemia.
British journal of haematology (2012-12-21)
John H Fargo, Christian P Kratz, Neelam Giri, Sharon A Savage, Carolyn Wong, Karen Backer, Blanche P Alter, Bertil Glader
ZUSAMMENFASSUNG
Diamond-Blackfan anaemia (DBA) is an inherited bone marrow failure syndrome (IBMFS) characterized by red cell aplasia. Mutations in ribosomal genes are found in more than 50% of cases. Elevated erythrocyte adenosine deaminase (eADA) was first noted in DBA in 1983. In this study we determined the value of eADA for the diagnosis of DBA compared with other IBMFS; the association of eADA in DBA with age, gender or other haematological parameters; and the association with known DBA-related gene mutations. For the diagnosis of DBA compared with non-DBA patients with other bone marrow failure syndromes, eADA had a sensitivity of 84%, specificity 95%, and positive and negative predictive values of 91%. In patients with DBA there was no association between eADA and gender, age, or other haematological parameters. Erythrocyte ADA segregated with, as well as independent of, known DBA gene mutations. While eADA was an excellent confirmatory test for DBA, 16% of patients with classical clinical DBA had a normal eADA.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type IX, ammonium sulfate suspension, 150-200 units/mg protein
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type X, buffered aqueous glycerol solution, ≥130 units/mg protein