Merck
  • Effect of enzymatic protein deamidation on protein solubility and flavor binding properties of soymilk.
Effect of enzymatic protein deamidation on protein solubility and flavor binding properties of soymilk.
Journal of food science (2013-01-03)
Inthawoot Suppavorasatit, Soo-Yeun Lee, Keith R Cadwallader
ZUSAMMENFASSUNG
The effect of enzymatic deamidation by protein-glutaminase (PG) on protein solubility and flavor binding potential of soymilk was studied. Treatment of soymilk with PG for 2 h (temperature of 44 °C and enzyme:substrate ratio (E/S) of 40 U/g protein) resulted in high degree of protein deamidation (66.4% DD) and relatively low degree of protein hydrolysis (4.25% DH). Deamidated (DSM) and control soymilks (CSM) did not differ with respect to aroma, but differed in taste characteristics by sensory evaluation. Protein solubility in DSM was enhanced at weakly acidic conditions (pH 5.0), but did not differ from non-deamidated soymilk at pH values of 3.0 and 7.0. Odor detection thresholds for the flavor compounds vanillin and maltol were approximately 5 and 3 fold lower, respectively, in DSM than in CSM. Dose-response curves (Fechner's law plots and n exponents from Stevens's power law) further demonstrated that DSM had a lower flavor binding potential than CSM. PG deamidation has the potential to reduce flavor binding problems encountered in high protein-containing foods and beverages. The findings of this study can help lead to the development of technology to produce protein-containing foods with improved functional properties, especially protein solubility, and potentially decreased flavor fade problems associated with flavor-protein interactions, especially with carbonyl containing flavor compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Vanillin, ReagentPlus®, 99%
Supelco
Vanillin Schmelzpunktstandard, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillin, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, ≥97%, FCC, FG
Sigma-Aldrich
Vanillin, tested according to Ph. Eur.
Supelco
Vanillin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glutaminase aus E. coli, Grade V, lyophilized powder, ≥50 units/mg protein
Supelco
Kalibriersubstanz von Mettler Toledo, ME 51143093, Vanillin, traceable to primary standards (LGC)
Sigma-Aldrich
Glutaminase aus E. coli, Grade VII, lyophilized powder, 500-1,500 units/mg protein
Vanillin, European Pharmacopoeia (EP) Reference Standard