Merck
Effect of molecular crowding on the temperature-pressure stability diagram of ribonuclease A.
Chemphyschem : a European journal of chemical physics and physical chemistry (2013-01-03)
Yong Zhai, Roland Winter
ZUSAMMENFASSUNG
FT-IR spectroscopic and thermodynamic measurements were designed to explore the effect of a macromolecular crowder, dextran, on the temperature and pressure-dependent phase diagram of the protein Ribonuclease A (RNase A), and we compare the experimental data with approximate theoretical predictions based on configuration entropy. Exploring the crowding effect on the pressure-induced unfolding of proteins provides insight in protein stability and folding under cell-like dense conditions, since pressure is a fundamental thermodynamic variable linked to molecular volume. Moreover, these studies are of relevance for understanding protein stability in deep-sea organisms, which have to cope with pressures in the kbar range. We found that not only temperature-induced equilibrium unfolding of RNase A, but also unfolding induced by pressure is markedly prohibited in the crowded dextran solutions, suggesting that crowded environments such as those found intracellularly, will also oppress high-pressure protein unfolding. The FT-IR spectroscopic measurements revealed a marked increase in unfolding pressure of 2 kbar in the presence of 30 wt % dextran. Whereas the structural changes upon thermal unfolding of the protein are not significantly influenced in the presence of the crowding agent, through stabilization by dextran the pressure-unfolded state of the protein retains more ordered secondary structure elements, which seems to be a manifestation of the entropic destabilization of the unfolded state by crowding.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type I-A, powder, ≥60% RNase A basis (SDS-PAGE), ≥50 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Molecular Biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, (Solution of 50% glycerol, 10mM Tris-HCL pH 8.0)
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type I-AS, 50-100 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type III-A, ≥85% RNase A basis (SDS-PAGE), 85-140 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type XII-A, ≥90% (SDS-PAGE), 75-125 Kunitz units/mg protein
Sigma-Aldrich
Ribonuklease B aus Rinderpankreas, BioReagent, ≥50 Kunitz units/mg protein, ≥80% (SDS-PAGE)
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type X-A, ≥90% (SDS-PAGE), ≥70 Kunitz units/mg protein