Merck
Recoding RNA editing of AZIN1 predisposes to hepatocellular carcinoma.
Nature medicine (2013-01-08)
Leilei Chen, Yan Li, Chi Ho Lin, Tim Hon Man Chan, Raymond Kwok Kei Chow, Yangyang Song, Ming Liu, Yun-Fei Yuan, Li Fu, Kar Lok Kong, Lihua Qi, Yan Li, Na Zhang, Amy Hin Yan Tong, Dora Lai-Wan Kwong, Kwan Man, Chung Mau Lo, Si Lok, Daniel G Tenen, Xin-Yuan Guan
ZUSAMMENFASSUNG
A better understanding of human hepatocellular carcinoma (HCC) pathogenesis at the molecular level will facilitate the discovery of tumor-initiating events. Transcriptome sequencing revealed that adenosine-to-inosine (A→I) RNA editing of AZIN1 (encoding antizyme inhibitor 1) is increased in HCC specimens. A→I editing of AZIN1 transcripts, specifically regulated by ADAR1 (encoding adenosine deaminase acting on RNA-1), results in a serine-to-glycine substitution at residue 367 of AZIN1, located in β-strand 15 (β15) and predicted to cause a conformational change, induced a cytoplasmic-to-nuclear translocation and conferred gain-of-function phenotypes that were manifested by augmented tumor-initiating potential and more aggressive behavior. Compared with wild-type AZIN1 protein, the edited form has a stronger affinity to antizyme, and the resultant higher AZIN1 protein stability promotes cell proliferation through the neutralization of antizyme-mediated degradation of ornithine decarboxylase (ODC) and cyclin D1 (CCND1). Collectively, A→I RNA editing of AZIN1 may be a potential driver in the pathogenesis of human cancers, particularly HCC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type IX, ammonium sulfate suspension, 150-200 units/mg protein
Sigma-Aldrich
Adenosine Deaminase from bovine spleen, Type X, buffered aqueous glycerol solution, ≥130 units/mg protein
Sigma-Aldrich
Anti-ADARB1 antibody produced in mouse, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-OAZ1 antibody produced in rabbit, affinity isolated antibody