Merck
  • A novel synthesized tyrosinase inhibitor: (E)-2-((2,4-dihydroxyphenyl)diazenyl)phenyl 4-methylbenzenesulfonate as an azo-resveratrol analog.
A novel synthesized tyrosinase inhibitor: (E)-2-((2,4-dihydroxyphenyl)diazenyl)phenyl 4-methylbenzenesulfonate as an azo-resveratrol analog.
Bioscience, biotechnology, and biochemistry (2013-01-08)
Sung Jin Bae, Young Mi Ha, Jin-Ah Kim, Ji Young Park, Tae Kwun Ha, Daeui Park, Pusoon Chun, Nam Hee Park, Hyung Ryong Moon, Hae Young Chung
ZUSAMMENFASSUNG
We synthesized a novel series of (E)-2-((substituted phenyl)diazenyl)phenyl 4-methylbenzenesulfonate derivatives (2 and 3) and (E)-2-((substituted phenyl)diazenyl)phenol derivatives (4 and 5), and conducted an evaluation in order to determine their inhibitory effects on mushroom tyrosinase, with the aim of discovering a tyrosinase inhibitor. Most of the compounds (3-5) exhibited higher inhibitory effects than kojic acid (IC(50) = 49.08 µM), a representative tyrosinase inhibitor. A novel synthesized compound, (E)-2-((2,4-dihydroxyphenyl)diazenyl)phenyl 4-methylbenzenesulfonate (3), showed the best results with an IC(50) of 17.85 µM, and showed competitive inhibition on Lineweaver-Burk plots, as further confirmed by the docking results. In addition, active compounds 3-5 were not cytotoxic to cultured B16F10 cells at the concentrations tested, and inhibited both tyrosinase and melanin synthesis. Therefore the active compounds (3-5) might be considered excellent candidates for use in the development of therapeutic agents for diseases associated with hyperpigmentation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyrosinase aus Ständerpilz, lyophilized powder, ≥1000 unit/mg solid
Sigma-Aldrich
Kojisäure