Merck
  • Separation of the antioxidant compound quercitrin from Lindera obtusiloba Blume and its antimelanogenic effect on B16F10 melanoma cells.
Separation of the antioxidant compound quercitrin from Lindera obtusiloba Blume and its antimelanogenic effect on B16F10 melanoma cells.
Bioscience, biotechnology, and biochemistry (2013-01-08)
Chung-Oui Hong, Hyun Ah Lee, Chae Hong Rhee, Se-Young Choung, Kwang-Won Lee
ZUSAMMENFASSUNG
Considering the growing evidence of the presence of antioxidant compounds in plant extracts, the objectives of this study were to identify antioxidant compounds in Lindera obtusiloba Blume (Lauraceae) and to evaluate their antimelanogenic activities on B16F10 melanoma cells. Organic solvent fractions were separated from L. obtusiloba extracts (LOE). The ethyl acetate fraction (LOE-E) was significantly active against oxidative damage induced by tert-butyl hydroperoxide in primary rat hepatocytes. Two single purified compounds, quercitrin (quercetin-3-O-α-L-rhamnopyranoside) and afzelin (kaempferol-3-O-α-L-rhamnoside), were identified by HPLC and NMR. These compounds were evaluated for antioxidant activities by 1,1-diphenyl 2-picrylhydrazyl (DPPH) radical scavenging assay and ferric reducing antioxidant power (FRAP) assay, and for their antimelanogenic activities by tyrosinase inhibitory assay melanin formation inhibition assay and Western bolt analysis for the signaling pathway. The significant effects of quercitrin on antioxidant and antimelanogenic activities, and signal modulation of ERK and MITF in B16F10 melanoma cells were observed. This is the first report to identify quercitrin in L. obtusiloba and its whitening effect.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.7%
Sigma-Aldrich
Ethylacetat, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
Tyrosinase aus Ständerpilz, lyophilized powder, ≥1000 unit/mg solid
Sigma-Aldrich
tert-Butylhydroperoxid -Lösung, 70 wt. % in H2O
Sigma-Aldrich
Ethylacetat, suitable for HPLC, ≥99.8%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
tert-Butylhydroperoxid -Lösung, 5.0-6.0 M in decane
Supelco
Ethylacetat, analytical standard
Sigma-Aldrich
tert-Butylhydroperoxid -Lösung, 5.0-6.0 M in nonane
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, biotech. grade, ≥99.8%
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Quercitrin, primary reference standard