Merck
  • The role of aldehyde/alcohol dehydrogenase (AdhE) in ethanol production from glycerol by Klebsiella pneumoniae.
The role of aldehyde/alcohol dehydrogenase (AdhE) in ethanol production from glycerol by Klebsiella pneumoniae.
Journal of industrial microbiology & biotechnology (2013-01-09)
Baek-Rock Oh, Won-Kyung Hong, Sun-Yeon Heo, Min-ho Joe, Jeong-Woo Seo, Chul Ho Kim
ZUSAMMENFASSUNG
Transcriptome analysis of a K. pneumoniae GEM167 mutant strain derived by irradiation with gamma rays, which exhibited high-level production of ethanol from glycerol, showed that the mutant expressed AdhE at a high level. Ethanol production decreased significantly, from 8.8 to 0.5 g l(-1), when an adhE-deficient derivative of that strain was grown on glycerol. Bacterial growth was also reduced under such conditions, showing that AdhE plays a critical role in maintenance of redox balance by catalyzing ethanol production. Overexpression of AdhE enhanced ethanol production, from pure or crude glycerol, to a maximal level of 31.9 g l(-1) under fed-batch fermentation conditions; this is the highest level of ethanol production from glycerol reported to date.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Alcohol-Dehydrogenase aus Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Alcohol-Dehydrogenase aus Saccharomyces cerevisiae, powder, ≥300 units/mg protein, mol wt ~141,000 (four subunits)
Sigma-Aldrich
Alcohol-Dehydrogenase aus Saccharomyces cerevisiae
Sigma-Aldrich
Alkoholdehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg