Merck
  • Vimentin is a component of a complex that binds to the 5'-UTR of human heme-regulated eIF2α kinase mRNA and regulates its translation.
Vimentin is a component of a complex that binds to the 5'-UTR of human heme-regulated eIF2α kinase mRNA and regulates its translation.
FEBS letters (2013-01-30)
Sangeeta Chatterjee, Amaresh C Panda, Sunil K Berwal, R K Sreejith, Charu Ritvika, Vasudevan Seshadri, Jayanta K Pal
ZUSAMMENFASSUNG
The human heme-regulated eIF2α kinase, also called the human heme-regulated inhibitor (hHRI) is significantly up-regulated particularly at the level of translation during stress. In this report we show that during lead-stress, the regulation of hHRI mRNA translation is mediated through its 5'-untranslated region (UTR) that interacts with specific trans-acting factors. Further, vimentin has been identified as one of the trans-acting factors that contribute to this regulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Blei(II)-acetat Trihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Blei(II)-acetat Trihydrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-102.0%
Sigma-Aldrich
Blei(II)-acetat Trihydrat, 99.999% trace metals basis
Sigma-Aldrich
Blei(II)-acetat Trihydrat, ≥99.99% trace metals basis