Merck
Optimizing asparaginase therapy for acute lymphoblastic leukemia.
Current opinion in oncology (2013-02-13)
Carmelo Rizzari, Valentino Conter, Jan Starý, Antonella Colombini, Anja Moericke, Martin Schrappe
ZUSAMMENFASSUNG
Asparaginases are important agents used in the treatment of children with acute lymphoblastic leukemia (ALL). Three types of asparaginase are currently available: two are derived from Escherichia coli [native asparaginase and pegylated asparaginase (PEG-asparaginase)] and one from Erwinia chrysanthemi (crisantaspase). All three products share the same mechanism of action but have different pharmacokinetic properties, which do not make them easily interchangeable. Among the known toxicities and side-effects, allergic reactions and silent inactivation represent the most important limitations to the prolonged use of any asparaginase product, with associated reduced therapeutic effects and poorer outcomes. Routine real time monitoring can help to identify patients with silent inactivation and facilitate a switch to a different product to ensure continued depletion of asparagine, completion of the treatment schedule and maintenance of outcomes. However, the most appropriate second-line treatment is still a matter of debate. PEG-asparaginase has lower immunogenicity and a longer half-life than native Escherichia coli (E. coli) asparaginase, which makes it useful for both first-line and second-line use with a reduced number of doses. However, PEG-asparaginase displays cross-reactivity with native E. coli asparaginase that may harm its therapeutic effects. Crisantaspase does not display cross-reactivity to either of the E. coli-derived products, which has made crisantaspase the second-line treatment option in a number of recent protocols. As crisantaspase has a much shorter biological half-life than the E. coli-derived products, the appropriate dosage and administration schedule are of paramount importance in delivering treatment with this product. In the ongoing trial AIEOP-BFM ALL 2009 (Associazione Italiana Ematologia Oncologia Pediatrica - Berlin-Franklin-Munster), in which PEG-asparaginase is used first-line, one dose of PEG-asparaginase is substituted by seven doses of crisantaspase given intravenously at 20,000 IU/m2 on alternate days when clinical allergy or silent inactivation is present. Based on the indications of different protocols, lack of cross-reactivity to the E. coli-derived products and taking into consideration regulatory factors and availability, crisantaspase may be considered a viable second-line therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Asparaginase aus E. coli, lyophilized powder, 100-300 units/mg protein (biuret)