Merck
Human PXR modulates hepatotoxicity associated with rifampicin and isoniazid co-therapy.
Nature medicine (2013-03-12)
Feng Li, Jie Lu, Jie Cheng, Laiyou Wang, Tsutomu Matsubara, Iván L Csanaky, Curtis D Klaassen, Frank J Gonzalez, Xiaochao Ma
ZUSAMMENFASSUNG
Co-therapy with rifampicin (RIF) and isoniazid (INH) used to treat tuberculosis in humans frequently causes liver injury. Here, using a pregnane X receptor (PXR)-humanized mouse model, we found that co-treatment with RIF and INH causes accumulation of the endogenous hepatotoxin protoporphyrin IX in the liver through PXR-mediated alteration of the heme biosynthesis pathway. These results provide insight into the mechanism of liver injury induced by co-treatment with these compounds and may lead to their safer use in the clinic.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous solution, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, lyophilized powder, ≥10 DEA units/mg solid
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, BioUltra, ≥5,700 DEA units/mg protein
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Acethydrazid, 90%
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥2,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, lyophilized powder, 30-60 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, ≥5,500 DEA units/mg protein
Sigma-Aldrich
Protoporphyrin IX Dinatriumsalz, ≥90% (HPLC)
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, buffered aqueous glycerol solution, 20-50 units/mg protein (in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Rinderdarmschleimhaut, buffered aqueous glycerol solution, ≥4,000 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus E. coli, ammonium sulfate suspension, 30-90 units/mg protein (modified Warburg-Christian, in glycine buffer)
Sigma-Aldrich
Phosphatase, alkalisch aus Schweineniere, lyophilized powder, ≥100 DEA units/mg protein
Sigma-Aldrich
Phosphatase, alkalisch aus Kälberdarmschleimhaut, suitable for enzyme immunoassay, solution (clear, colorless), ~2500 U/mg protein (~10 mg/ml)
Sigma-Aldrich
Phosphatase, alkalische Garnelen, ≥900 DEA units/mL, buffered aqueous glycerol solution, recombinant, expressed in proprietary host
Sigma-Aldrich
Phosphatase, alkalisch Rind, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein